Learn More
Biomatik Corporation Recombinant Human Bcl2 Associated X Protein (Bax)
Supplier:  Biomatik Corporation RPU55279
Encompass_Preferred
Recombinant Human Bcl2 Associated X Protein (Bax) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Bcl2 Associated X Protein (Bax). Target Synonyms: BCL2L4; Bax Zeta; Apoptosis regulator BAX; Bcl-2-like protein 4. Accession Number: Q07812. Gene ID: 581. Expression Region: Met1~Gln171. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0286