Learn More
Biomatik Corporation Recombinant Human Adropin (AD)
Supplier:  Biomatik Corporation RPU55406
Encompass_Preferred
Recombinant Human Adropin (AD) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Adropin (AD). Target Synonyms: ENHO; UNQ470; Energy Homeostasis Associated Protein. Accession Number: Q6UWT2. Gene ID: 375704. Expression Region: Cys34~Pro76. Tag Info: Two N-terminal Tags, His-tag and SUMO-tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0293