Learn More
Biomatik Corporation Recombinant Rat Interleukin 11 (IL11)
Supplier:  Biomatik Corporation RPU55464
Encompass_Preferred
Recombinant Rat Interleukin 11 (IL11) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Interleukin 11 (IL11). Target Synonyms: AGIF; Adipogenesis Inhibitory Factor; Oprelvekin. Accession Number: Q99MF5. Expression Region: Pro25~Leu199. Tag Info: N-terminal His Tag. Theoretical MW: 23kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0296