Learn More
Biomatik Corporation Human Matrix Metalloproteinase 13 (MMP13), Active Protein
Supplier:  Biomatik Corporation RPU55483
Encompass_Preferred
Human Matrix Metalloproteinase 13 (MMP13), Active Protein is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Matrix Metalloproteinase 13 (MMP13). Target Synonyms: CLG3; Collagenase 3. Accession Number: P45452. Gene ID: 4322. Expression Region: Glu103~Leu290. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0301