Learn More
Biomatik Corporation Human Perforin 1 (PRF1), Active Protein
Supplier:  Biomatik Corporation RPU55672
Encompass_Preferred
Human Perforin 1 (PRF1), Active Protein is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Perforin 1 (PRF1). Target Synonyms: FLH2; HPLH2; P1; PFP; Pore Forming Protein; Cytolysin; Lymphocyte pore-forming protein. Accession Number: P14222. Gene ID: 5551. Expression Region: Lys32~Phe316. Tag Info: N-terminal His and GST Tag. Theoretical MW: 62kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0312