Learn More
Biomatik Corporation Recombinant Ecotin (ECO)
Supplier:  Biomatik Corporation RPU41317
Encompass_Preferred
Recombinant Ecotin (ECO) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Universal. Target Name: Ecotin (ECO). Accession Number: P23827. Gene ID: 946700. Expression Region: Ala21~Arg162. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9938