Learn More
Biomatik Corporation Mouse Tissue Inhibitors Of Metalloproteinase 3 (TIMP3), Active Protein
Supplier:  Biomatik Corporation RPU56338
Encompass_Preferred
Mouse Tissue Inhibitors Of Metalloproteinase 3 (TIMP3), Active Protein is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Tissue Inhibitors Of Metalloproteinase 3 (TIMP3). Target Synonyms: SFD; TIMP Metallopeptidase Inhibitor 3; Metallopeptidase Inhibitor 3; Sorsby Fundus Dystrophy; Pseudoinflammatory. Accession Number: P39876. Gene ID: 21859. Expression Region: Gly20~Pro211. Tag Info: N-terminal His Tag. Theoretical MW: 28/22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0337