Learn More
Biomatik Corporation Native Mouse Fibrinogen (FG)
Supplier:  Biomatik Corporation RPU56375
Encompass_Preferred
Native Mouse Fibrinogen (FG) is a purified Natural Protein. Purity: >90% by SDS-PAGE. Host: Mouse. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Fibrinogen (FG). Target Synonyms: F1; FI; Fbg; Coagulation Factor I. Accession Number: E9PV24; Q8K0E8; Q8VCM7. Theoretical MW: 70/60/55kDa. Buffer: PBS, pH7.4, containing 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0339