Learn More
Biomatik Corporation Recombinant Rat Fibroblast Growth Factor 4 (FGF4)
Supplier:  Biomatik Corporation RPU59431
Encompass_Preferred
Recombinant Rat Fibroblast Growth Factor 4 (FGF4) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Fibroblast Growth Factor 4 (FGF4). Target Synonyms: HBGF4; HST; HSTF1; KFGF; K-FGF; Heparin Secretory Transforming Protein 1; Kaposi Sarcoma Oncogene; Transforming Protein KS3; FGF-Related Oncogene. Accession Number: Q8R5L6. Expression Region: Ser67~Leu195. Tag Info: N-terminal His and GST Tag. Theoretical MW: 45kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0361