Learn More
Biomatik Corporation Recombinant Escherichia coli DNA mismatch repair protein MutL (mutL)
Supplier:  Biomatik Corporation RPC20146
Encompass_Preferred
Recombinant Escherichia coli DNA mismatch repair protein MutL (mutL) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12). Target Name: mutL. Target Synonyms: mutL; b4170; JW4128; DNA mismatch repair protein MutL. Accession Number: P23367. Expression Region: 1~615aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 71.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0381