Learn More
Biomatik Corporation Recombinant Human Fibrinogen Like Protein 1 (FGL1)
Supplier:  Biomatik Corporation RPU41423
Encompass_Preferred
Recombinant Human Fibrinogen Like Protein 1 (FGL1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Fibrinogen Like Protein 1 (FGL1). Target Synonyms: FGL-1; HFREP1; HP-041; LFIRE1; Hepassocin; Hepatocyte-derived fibrinogen-related protein 1; Liver fibrinogen-related protein 1. Accession Number: Q08830. Gene ID: 2267. Expression Region: Leu23~Ile312. Tag Info: N-terminal His Tag. Theoretical MW: 41kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9942