Learn More
Biomatik Corporation Recombinant Glycine max Stress-induced protein SAM22 (PR-10)
Supplier:  Biomatik Corporation RPC20254
Encompass_Preferred
Recombinant Glycine max Stress-induced protein SAM22 (PR-10) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Glycine max (Soybean) (Glycine hispida). Target Name: PR-10. Target Synonyms: PR-10; GLYMA_07G243500; Stress-induced protein SAM22; Pathogenesis-related protein 10; Starvation-associated message 22; allergen Gly m 4. Accession Number: P26987. Expression Region: 1~158aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 18.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0392