Learn More
Biomatik Corporation Recombinant Arachis hypogaea Allergen Ara h 1, clone P17
Supplier:  Biomatik Corporation RPC20274
Encompass_Preferred
Recombinant Arachis hypogaea Allergen Ara h 1, clone P17 is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Arachis hypogaea (Peanut). Target Name: Arachis hypogaea Allergen Ara h 1, clone P17. Target Synonyms: Allergen Ara h IAllergen: Ara h 1. Accession Number: P43237. Expression Region: 26~216aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 39kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0396