Learn More
Biomatik Corporation Recombinant Betula pendula Major pollen allergen Bet v 1-A (BETVIA)
Supplier:  Biomatik Corporation RPC20315
Encompass_Preferred
Recombinant Betula pendula Major pollen allergen Bet v 1-A (BETVIA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Betula pendula (European white birch) (Betula verrucosa). Target Name: BETVIA. Target Synonyms: Allergen Bet v I-AAllergen: Bet v 1-A. Accession Number: P15494. Expression Region: 2~160aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 33.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0403