Learn More
Biomatik Corporation Recombinant Pseudomonas aeruginosa Protease lasA (lasA)
Supplier:  Biomatik Corporation RPC20467
Encompass_Preferred
Recombinant Pseudomonas aeruginosa Protease lasA (lasA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228). Target Name: lasA. Target Synonyms: Staphylolytic protease. Accession Number: P14789. Expression Region: 237~418aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 36kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0412