Learn More
Biomatik Corporation Recombinant Mouse Dihydrolipoyllysine-residue acetyltransferase component of pyruvate dehydrogenase complex, mitochondrial (Dlat)
Supplier:  Biomatik Corporation RPC20601
Encompass_Preferred
Recombinant Mouse Dihydrolipoyllysine-residue acetyltransferase component of pyruvate dehydrogenase complex, mitochondrial (Dlat) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Dlat. Target Synonyms: Dihydrolipoamide acetyltransferase component of pyruvate dehydrogenase complexPyruvate dehydrogenase complex component E2. Accession Number: Q8BMF4. Expression Region: 86~642aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 62.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0415