Learn More
Biomatik Corporation Recombinant Rabbit Fibroblast Growth Factor 2, Basic (FGF2)
Supplier:  Biomatik Corporation RPU41441
Encompass_Preferred
Recombinant Rabbit Fibroblast Growth Factor 2, Basic (FGF2) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rabbit (Oryctolagus cuniculus). Target Name: Fibroblast Growth Factor 2, Basic (FGF2). Target Synonyms: B-FGF; BFGF; FGFB; HBGH-2; Basic Fibroblast Growth Factor; Heparin-binding growth factor 2. Accession Number: P48799. Expression Region: Phe26~Ala153. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9946