Learn More
Biomatik Corporation Recombinant Human Protein cereblon (CRBN)
Supplier:  Biomatik Corporation RPC20606
Encompass_Preferred
Recombinant Human Protein cereblon (CRBN) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: In Vitro E. coli Expression System. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CRBN. Target Synonyms: 2610203G15Rik; 2900045O07Rik; AF229032; AW108261; Cereblon; Crbn; CRBN_HUMAN; DKFZp781K0715; MGC27358; MRT2A; OTTHUMP00000209555; piL; Protein cereblon; Protein x 0001. Accession Number: Q96SW2. Expression Region: 1~442aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 64.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0417