Learn More
Biomatik Corporation Recombinant Human Claudin-6 (CLDN6), partial
Supplier:  Biomatik Corporation RPC20717
Encompass_Preferred
Recombinant Human Claudin-6 (CLDN6), partial is a purified CF Transmembrane Protein. Purity: >90% as determined by SDS-PAGE. Host: In Vitro E. coli Expression System. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CLDN6. Target Synonyms: Skullin. Accession Number: P56747. Expression Region: 1~82aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 24.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0422