Learn More
Biomatik Corporation Recombinant Human Asporin (ASPN)
Supplier:  Biomatik Corporation RPC20946
Encompass_Preferred
Recombinant Human Asporin (ASPN) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ASPN. Target Synonyms: Periodontal ligament-associated protein 1PLAP-1. Accession Number: Q9BXN1. Expression Region: 33~380aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 55.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0434