Learn More
Biomatik Corporation Recombinant Human CD44 antigen (CD44), partial
Supplier:  Biomatik Corporation RPC21057
Encompass_Preferred
Recombinant Human CD44 antigen (CD44), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CD44. Target Synonyms: CDw44Epican; Extracellular domain matrix receptor III; ECMR-IIIGP90 lymphocyte homing/adhesion receptor; HUTCH-IHeparan sulfate proteoglycan; Hermes antigen; Hyaluronate receptor; Phagocytic glycoprotein 1; PGP-1; Phagocytic glycoprotein I; PGP-I; CD44. Accession Number: P16070. Expression Region: 21~606aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 80.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0447