Learn More
Biomatik Corporation Recombinant Human Muscarinic acetylcholine receptor M3 (CHRM3), partial
Supplier:  Biomatik Corporation RPC21095
Encompass_Preferred
Recombinant Human Muscarinic acetylcholine receptor M3 (CHRM3), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CHRM3. Target Synonyms: AChR; ACM3_HUMAN; cholinergic receptor muscarinic 3; CHRM 3; CHRM3; EGBRS; HM 3; HM3; m3 muscarinic acetylcholine receptor; M3 muscarinic receptor; muscarinic 3; Muscarinic acetylcholine receptor M3; muscarinic cholinergic m3 receptor; muscarinic m3 receptor. Accession Number: P20309. Expression Region: 253~492aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 40.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0449