Learn More
Biomatik Corporation Recombinant Mouse Ficolin 1 (FCN1)
Supplier:  Biomatik Corporation RPU41491
Encompass_Preferred
Recombinant Mouse Ficolin 1 (FCN1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Ficolin 1 (FCN1). Target Synonyms: FCNM; Collagen/fibrinogen domain-containing protein 1; Ficolin-alpha; Ficolin-A; M-ficolin. Accession Number: O70165. Gene ID: 14133. Expression Region: Arg25~Gly317. Tag Info: N-terminal His Tag. Theoretical MW: 35kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9948