Learn More
Biomatik Corporation Recombinant Human Receptor tyrosine-protein kinase erbB-2 (ERBB2), partial
Supplier:  Biomatik Corporation RPC21276
Encompass_Preferred
Recombinant Human Receptor tyrosine-protein kinase erbB-2 (ERBB2), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ERBB2. Target Synonyms: Metastatic lymph node gene 19 protein; MLN 19Proto-oncogene NeuProto-oncogene c-ErbB-2Tyrosine kinase-type cell surface receptor HER2p185erbB2;; CD340. Accession Number: P04626. Expression Region: 23~652aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 73.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0472