Learn More
Biomatik Corporation Recombinant Human Prothrombin (F2)
Supplier:  Biomatik Corporation RPC21291
Encompass_Preferred
Recombinant Human Prothrombin (F2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: F2. Target Synonyms: Coagulation factor IICleaved into the following 4 chains:Activation peptide fragment 1Activation peptide fragment 2Thrombin light chainThrombin heavy chain. Accession Number: P00734. Expression Region: 44~622aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 81.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0473