Learn More
Biomatik Corporation Recombinant Human Fibrinogen beta chain (FGB), partial
Supplier:  Biomatik Corporation RPC21313
Encompass_Preferred
Recombinant Human Fibrinogen beta chain (FGB), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: FGB. Target Synonyms: Beta fibrinogen; Epididymis Secretory Sperm Binding Protein Li 78p; FGB; FIBB_HUMAN; Fibrinogen beta chain; Fibrinogen; B Beta Polypeptide; HEL-S-78p. Accession Number: P02675. Expression Region: 45~491aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 54.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0477