Learn More
Biomatik Corporation Recombinant Mouse Interleukin-2 receptor subunit beta (Il2rb), partial
Supplier:  Biomatik Corporation RPC21516
Encompass_Preferred
Recombinant Mouse Interleukin-2 receptor subunit beta (Il2rb), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: l2rb. Target Synonyms: High affinity IL-2 receptor subunit betap70-75; CD122. Accession Number: P16297. Expression Region: 24~240aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 29.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0496