Learn More
Biomatik Corporation Recombinant Rat Galectin-3 (Lgals3)
Supplier:  Biomatik Corporation RPC21595
Encompass_Preferred
Recombinant Rat Galectin-3 (Lgals3) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Lgals3. Target Synonyms: 35 kDa lectinCarbohydrate-binding protein 35. Accession Number: P08699. Expression Region: 2~262aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 31.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0503