Learn More
Biomatik Corporation Recombinant Human Apolipoprotein M (APOM)
Supplier:  Biomatik Corporation RPC20921
Encompass_Preferred
Recombinant Human Apolipoprotein M (APOM) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: APOM. Target Synonyms: Protein G3a. Accession Number: O95445. Expression Region: 1~188aa. Tag Info: N-Terminal 10Xhis-Gst-Tagged. Theoretical MW: 49.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0432