Learn More
Biomatik Corporation Recombinant Human Carcinoembryonic antigen-related cell adhesion molecule 6 (CEACAM6)
Supplier:  Biomatik Corporation RPC21080
Encompass_Preferred
Recombinant Human Carcinoembryonic antigen-related cell adhesion molecule 6 (CEACAM6) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CEACAM6. Target Synonyms: Non-specific crossreacting antigen; Normal cross-reacting antigen; CD_antigen: CD66c. Accession Number: P40199. Expression Region: 35~320aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 47.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0448