Learn More
Biomatik Corporation Recombinant Human Glial Cell Line Derived Neurotrophic Factor (GDNF)
Supplier:  Biomatik Corporation RPU41604
Encompass_Preferred
Recombinant Human Glial Cell Line Derived Neurotrophic Factor (GDNF) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Glial Cell Line Derived Neurotrophic Factor (GDNF). Target Synonyms: ATF1; ATF2; HFB1-GDNF; Astrocyte-Derived Trophic Factor; Glial Derived Neurotrophic Factor. Accession Number: P39905. Gene ID: 2668. Expression Region: Ser78~Ile211. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9955