Learn More
Biomatik Corporation Recombinant Mouse Protein CYR61 (Cyr61)
Supplier:  Biomatik Corporation RPC21186
Encompass_Preferred
Recombinant Mouse Protein CYR61 (Cyr61) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Cyr61. Target Synonyms: 3CH61CCN family member 1Cysteine-rich angiogenic inducer 61Insulin-like growth factor-binding protein 10. Accession Number: P18406. Expression Region: 25~379aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 42.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0466