Learn More
Biomatik Corporation Recombinant Human Granzyme B (GZMB)
Supplier:  Biomatik Corporation RPC21426
Encompass_Preferred
Recombinant Human Granzyme B (GZMB) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: GZMB. Target Synonyms: C11CTLA-1Cathepsin G-like 1; CTSGL1Cytotoxic T-lymphocyte proteinase 2; Lymphocyte proteaseFragmentin-2Granzyme-2Human lymphocyte protein; HLPSECTT-cell serine protease 1-3E. Accession Number: P10144. Expression Region: 21~247aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 31.0kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0488