Learn More
Biomatik Corporation Recombinant Human Leucine-rich repeat-containing G-protein coupled receptor 5 (LGR5), partial
Supplier:  Biomatik Corporation RPC21603
Encompass_Preferred
Recombinant Human Leucine-rich repeat-containing G-protein coupled receptor 5 (LGR5), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: LGR5. Target Synonyms: G-protein coupled receptor 49G-protein coupled receptor 67G-protein coupled receptor HG38. Accession Number: O75473. Expression Region: 22~561aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 65.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0505