Learn More
Biomatik Corporation Recombinant Mouse Granzyme M (GZMM)
Supplier:  Biomatik Corporation RPU41737
Encompass_Preferred
Recombinant Mouse Granzyme M (GZMM) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Granzyme M (GZMM). Target Synonyms: GZM-M; LMET1; MET1; Hu-Met-1; Met-ase; Lymphocyte Met Ase 1; Met-1 serine protease; Natural killer cell granular protease. Accession Number: O08643. Gene ID: 16904. Expression Region: Arg31~Arg257. Tag Info: N-terminal His and GST Tag. Theoretical MW: 55kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9965