Learn More
Biomatik Corporation Recombinant Mouse Apolipoprotein M (APOM)
Supplier:  Biomatik Corporation RPU40367
Encompass_Preferred
Recombinant Mouse Apolipoprotein M (APOM) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Apolipoprotein M (APOM). Target Synonyms: Apo-M; G3a; NG20; Protein G3a. Accession Number: Q9Z1R3. Gene ID: 55938. Expression Region: Met20~Lys190. Tag Info: N-terminal His Tag. Theoretical MW: 32kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9884