Learn More
Biomatik Corporation Recombinant Pig Interferon Alpha (IFNa)
Supplier:  Biomatik Corporation RPU42108
Encompass_Preferred
Recombinant Pig Interferon Alpha (IFNa) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Pig (Sus scrofa; Porcine). Target Name: Interferon Alpha (IFNa). Target Synonyms: IFNA1; IFL; LeIF D; IFN; IFN-Alpha; IFNA13; IFN-A; IFNAP22; IFN-Alpha 1b; Interferon Alpha 1b; Interferon, Leukocytic; IFN, Leukocyte; Interferon alpha-D. Accession Number: P49879. Expression Region: Cys24~Met172. Tag Info: N-terminal His and GST Tag. Theoretical MW: 44kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9983
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'