Learn More
Biomatik Corporation Recombinant Human Interleukin 1 Zeta (IL1z)
Supplier:  Biomatik Corporation RPU42202
Encompass_Preferred
Recombinant Human Interleukin 1 Zeta (IL1z) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interleukin 1 Zeta (IL1z). Target Synonyms: IL1F7; IL37; FIL1; FIL1(ZETA); FIL1Z; IL-1F7; IL-1H4; IL-1RP1; IL1H4; IL1RP1; Interleukin 37; Interleukin-1-Related Protein; Interleukin 1 Family, Member 7. Accession Number: Q9NZH6. Gene ID: 27178. Expression Region: Lys27~Asp192. Tag Info: N-terminal His Tag. Theoretical MW: 22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9988