Learn More
Biomatik Corporation Recombinant Human Laminin Gamma 2 (LAMC2)
Supplier:  Biomatik Corporation RPU42480
Encompass_Preferred
Recombinant Human Laminin Gamma 2 (LAMC2) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Laminin Gamma 2 (LAMC2). Target Synonyms: B2T; EBR2; EBR2A; LAMB2T; LAMNB2; Kalinin/nicein/epiligrin 100 kDa subunit; Cell-scattering factor 140 kDa subunit; Ladsin 140 kDa; Large adhesive scatter factor 140 kDa. Accession Number: Q13753. Gene ID: 3918. Expression Region: Cys28~Cys196. Tag Info: N-terminal His Tag. Theoretical MW: 22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0003