Learn More
Biomatik Corporation Recombinant Human Bone Morphogenetic Protein 15 (BMP15)
Supplier:  Biomatik Corporation RPU40483
Encompass_Preferred
Recombinant Human Bone Morphogenetic Protein 15 (BMP15) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Bone Morphogenetic Protein 15 (BMP15). Target Synonyms: GDF9B; GDF9-B; ODG2; POF4; Growth/differentiation factor 9B. Accession Number: O95972. Gene ID: 9210. Expression Region: Thr266~Ser388. Tag Info: N-terminal His Tag. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9888