Learn More
Biomatik Corporation Recombinant Human Low Density Lipoprotein Receptor Related Protein 1 (LRP1)
Supplier:  Biomatik Corporation RPU42574
Encompass_Preferred
Recombinant Human Low Density Lipoprotein Receptor Related Protein 1 (LRP1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Low Density Lipoprotein Receptor Related Protein 1 (LRP1). Target Synonyms: CD91; LRP; A2MR; APOER; APR; TGFBR5; Alpha-2-Macroglobulin Receptor; Prolow-density lipoprotein receptor-related protein 1; Apolipoprotein E receptor. Accession Number: Q07954. Gene ID: 4035. Expression Region: Val4441~Ala4544. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0008