Learn More
Biomatik Corporation Recombinant Mouse Mucin 1 (MUC1)
Supplier:  Biomatik Corporation RPU42816
Encompass_Preferred
Recombinant Mouse Mucin 1 (MUC1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Mucin 1 (MUC1). Target Synonyms: CD227; EMA; H23AG; KL-6; PEM; PEMT; PUM; CA15-3; CA153; Cancer antigen 15-3; Carcinoma-associated mucin; Episialin; Peanut-reactive urinary mucin; Polymorphic epithelial mucin. Accession Number: Q02496. Gene ID: 17829. Expression Region: Ser410~Ala525. Tag Info: N-terminal His Tag. Theoretical MW: 10kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0016