Learn More
Biomatik Corporation Recombinant Human N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP)
Supplier:  Biomatik Corporation RPU43096
Encompass_Preferred
Recombinant Human N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP). Target Synonyms: NT-Pro-BNP;; N-BNP. Accession Number: P16860. Gene ID: 4879. Expression Region: His27~Arg102. Tag Info: N-terminal His Tag. Theoretical MW: 12kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0028