Learn More
Biomatik Corporation Recombinant Human Promyelocytic Leukemia Protein (PML)
Supplier:  Biomatik Corporation RPU43439
Encompass_Preferred
Recombinant Human Promyelocytic Leukemia Protein (PML) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Promyelocytic Leukemia Protein (PML). Target Synonyms: MYL; RNF71; TRIM19; Probable Transcription Factor PML; Ring Finger Protein 71; Tripartite motif-containing protein 19. Accession Number: P29590. Gene ID: 5371. Expression Region: Gln59~Glu239. Tag Info: N-terminal His Tag. Theoretical MW: 31kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0046