Learn More
Biomatik Corporation Recombinant Human Pyruvate kinase isozymes M2 (PKM2)
Supplier:  Biomatik Corporation RPU43549
Encompass_Preferred
Recombinant Human Pyruvate kinase isozymes M2 (PKM2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Pyruvate kinase isozymes M2 (PKM2). Target Synonyms: M2PK; PKM; M2-PK; PKM1; CTHBP; OIP3; PK3; PKM; TCB; THBP1; Pyruvate kinase muscle isozyme; Thyroid hormone-binding protein 1; Cytosolic thyroid hormone-binding protein; Pyruvate Kinase, Muscle. Accession Number: P14618. Gene ID: 5315. Expression Region: Arg294~Phe470. Tag Info: N-terminal His and GST Tag. Theoretical MW: 50kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0051