Learn More
Biomatik Corporation Recombinant Human Ribonuclease T2 (RNASET2)
Supplier:  Biomatik Corporation RPU43656
Encompass_Preferred
Recombinant Human Ribonuclease T2 (RNASET2) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Ribonuclease T2 (RNASET2). Target Synonyms: Rnase-T2; RNASE6PL; RNASE6-PL; Ribonuclease 6. Accession Number: O00584. Gene ID: 8635. Expression Region: Asp25~His256. Tag Info: N-terminal His and GST Tag. Theoretical MW: 57kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0053