Learn More
Biomatik Corporation Recombinant Rat S100 Calcium Binding Protein B (S100B)
Supplier:  Biomatik Corporation RPU43705
Encompass_Preferred
Recombinant Rat S100 Calcium Binding Protein B (S100B) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: S100 Calcium Binding Protein B (S100B). Target Synonyms: S100-B; NEF; S100Beta; S-100 protein subunit beta. Accession Number: P04631. Gene ID: 25742. Expression Region: Met1~Glu92. Tag Info: N-terminal His Tag. Theoretical MW: 12kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0055