Learn More
Biomatik Corporation Recombinant Rat T-Cell Immunoreceptor With Ig And ITIM Domains Protein (TIGIT)
Supplier:  Biomatik Corporation RPU43959
Encompass_Preferred
Recombinant Rat T-Cell Immunoreceptor With Ig And ITIM Domains Protein (TIGIT) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: T-Cell Immunoreceptor With Ig And ITIM Domains Protein (TIGIT). Target Synonyms: VSIG9; VSTM3; V-Set And Immunoglobulin Domain Containing 9; V-Set And Transmembrane Domain Containing 3. Accession Number: D3ZTQ2. Expression Region: Met28~Ser227. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0061