Learn More
Medchemexpress LLC Catalpol | 2415-24-9 | 100 MG
SDP
Catalog No. 5000417092
Encompass_Preferred
Catalpol (Catalpinoside) is an iridoid glycoside found in Rehmannia glutinosa. It exhibits neuroprotective, hypoglycemic, anti-inflammatory, anti-cancer, anti-spasmodic, anti-oxidant, and anti-HBV effects.
  • Neuroprotective effects
  • Hypoglycemic effects
  • Anti-inflammatory effects
  • Anti-cancer effects
  • Anti-spasmodic effects
  • Anti-oxidant effects
  • Anti-HBV effects
Catalog No. 50-004-17092 Supplier Medchemexpress LLC Supplier No. HYN0820100MG
Catalpol (Catalpinoside) is an iridoid glycoside found in Rehmannia glutinosa. It exhibits neuroprotective, hypoglycemic, anti-inflammatory, anti-cancer, anti-spasmodic, anti-oxidant, and anti-HBV effects.
  • Neuroprotective effects
  • Hypoglycemic effects
  • Anti-inflammatory effects
  • Anti-cancer effects
  • Anti-spasmodic effects
  • Anti-oxidant effects
  • Anti-HBV effects