Learn More
Medchemexpress LLC Catalpol | 2415-24-9 | 362.33 | 50 MG
SDP
Catalog No. 5000416238
Encompass_Preferred
Catalpol is an iridoid glycoside found in *Rehmannia glutinosa*. It exhibits various effects including neuroprotective, hypoglycemic, anti-inflammatory, anti-cancer, anti-spasmodic, anti-oxidant, and anti-HBV effects.
  • Found in *Rehmannia glutinosa*
  • Exhibits neuroprotective effects
  • Exhibits hypoglycemic effects
  • Exhibits anti-inflammatory effects
  • Exhibits anti-cancer effects
  • Exhibits anti-spasmodic effects
  • Exhibits anti-oxidant effects
  • Exhibits anti-HBV effects
  • Induces neuronal differentiation in PC12 cells
  • Promotes neurite length
  • Reduces blood urea nitrogen in renal ischemia/reperfusion injury (IRI) mice
  • Reduces serum creatinine levels in IRI mice
  • Reduces KIM-1 expression in IRI mice
  • Suppresses PI3K, Akt, eNOS levels
  • Suppresses TNF-α, IL-1β, IL-6, and IL-10 activities
Catalog No. 50-004-16238 Supplier Medchemexpress LLC Supplier No. HYN082050MG
Catalpol is an iridoid glycoside found in *Rehmannia glutinosa*. It exhibits various effects including neuroprotective, hypoglycemic, anti-inflammatory, anti-cancer, anti-spasmodic, anti-oxidant, and anti-HBV effects.
  • Found in *Rehmannia glutinosa*
  • Exhibits neuroprotective effects
  • Exhibits hypoglycemic effects
  • Exhibits anti-inflammatory effects
  • Exhibits anti-cancer effects
  • Exhibits anti-spasmodic effects
  • Exhibits anti-oxidant effects
  • Exhibits anti-HBV effects
  • Induces neuronal differentiation in PC12 cells
  • Promotes neurite length
  • Reduces blood urea nitrogen in renal ischemia/reperfusion injury (IRI) mice
  • Reduces serum creatinine levels in IRI mice
  • Reduces KIM-1 expression in IRI mice
  • Suppresses PI3K, Akt, eNOS levels
  • Suppresses TNF-α, IL-1β, IL-6, and IL-10 activities